Product Details

SNP ID
rs80138577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83243606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACCCAAAGGCACTGGCTTGAGA[C/T]GCTGGAGTCAGAACTATAGGTCCGT
Phenotype
MIM: 607619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRMD3 PubMed Links

Gene Details

Gene
FRMD3
Gene Name
FERM domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244959.1 Intron NP_001231888.1
NM_001244960.1 Intron NP_001231889.1
NM_001244961.1 Intron NP_001231890.1
NM_001244962.1 Intron NP_001231891.1
NM_174938.5 Intron NP_777598.3
XM_017014588.1 Intron XP_016870077.1
XM_017014589.1 Intron XP_016870078.1
XM_017014590.1 Intron XP_016870079.1
XM_017014591.1 Intron XP_016870080.1
Gene
LOC102723989
Gene Name
uncharacterized LOC102723989
There are no transcripts associated with this gene.

View Full Product Details