Product Details

SNP ID
rs78107299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:85551478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGTGTGCCTATATTCTCAGTA[C/T]TAAATCACAAGTGCCTTTAAGATCA
Phenotype
MIM: 606830
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGTPBP1 PubMed Links

Gene Details

Gene
AGTPBP1
Gene Name
ATP/GTP binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286715.1 Intron NP_001273644.1
NM_001286717.1 Intron NP_001273646.1
NM_015239.2 Intron NP_056054.2
XM_005251847.3 Intron XP_005251904.1
XM_005251848.2 Intron XP_005251905.1
XM_005251849.3 Intron XP_005251906.1
XM_011518418.2 Intron XP_011516720.1
XM_011518420.2 Intron XP_011516722.1
XM_011518421.2 Intron XP_011516723.1
XM_017014544.1 Intron XP_016870033.1
XM_017014545.1 Intron XP_016870034.1
XM_017014546.1 Intron XP_016870035.1
XM_017014547.1 Intron XP_016870036.1
XM_017014548.1 Intron XP_016870037.1
XM_017014549.1 Intron XP_016870038.1
XM_017014550.1 Intron XP_016870039.1

View Full Product Details