Product Details

SNP ID
rs75568275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97501192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAACTACGTGTAGACAAAACGAT[A/C]GCGGTTAAAGCTTGCGGCTCAGAGC
Phenotype
MIM: 611258 MIM: 190930
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TDRD7 PubMed Links

Gene Details

Gene
TDRD7
Gene Name
tudor domain containing 7
There are no transcripts associated with this gene.

Gene
TMOD1
Gene Name
tropomodulin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166116.1 13 UTR 5 NP_001159588.1
NM_003275.3 13 Intron NP_003266.1

View Full Product Details