Product Details

SNP ID
rs74908311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94578738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTAATAATAACATACCATTTCCT[G/T]TTGTGCTGTAGCAAAGATATAAATT
Phenotype
MIM: 616146
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TGDS PubMed Links

Gene Details

Gene
TGDS
Gene Name
TDP-glucose 4,6-dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304430.1 866 Missense Mutation AAA,AAC K185N NP_001291359.1
NM_014305.3 866 Missense Mutation AAA,AAC K217N NP_055120.1
XM_011521065.2 866 Missense Mutation AAA,AAC K185N XP_011519367.1
XM_011521066.2 866 Missense Mutation AAA,AAC K185N XP_011519368.1

View Full Product Details