Product Details

SNP ID
rs77451585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40600478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAAAGGAAGTTAAATTTTGATC[C/T]CCCATGGTGGGAAGACAGACAGTTT
Phenotype
MIM: 609173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KNL1 PubMed Links

Gene Details

Gene
KNL1
Gene Name
kinetochore scaffold 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144508.4 Intron NP_653091.3
NM_170589.4 Intron NP_733468.3
XM_017022432.1 Intron XP_016877921.1

View Full Product Details