Product Details

SNP ID
rs78142209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72398406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGGGGAACCGGAAATACCAAA[C/G]GGTGAGGAGGTATCTAATTGAAAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105370888 PubMed Links

Gene Details

Gene
LOC105370888
Gene Name
uncharacterized LOC105370888
There are no transcripts associated with this gene.

Gene
TMEM202
Gene Name
transmembrane protein 202
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080462.1 80 Missense Mutation ACG,AGG T27R NP_001073931.1
XM_011521497.1 80 UTR 5 XP_011519799.1
XM_011521499.1 80 UTR 5 XP_011519801.1

View Full Product Details