Product Details

SNP ID
rs78949160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89683669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTCTCTTTCTTTGCCCTGTCA[C/G]ATGTAACTCGTTTCATCTGCAGGGA
Phenotype
MIM: 603866 MIM: 170290
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PEX11A PubMed Links

Gene Details

Gene
PEX11A
Gene Name
peroxisomal biogenesis factor 11 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271572.1 488 Missense Mutation TCT,TGT S120C NP_001258501.1
NM_001271573.1 488 Missense Mutation TCT,TGT S6C NP_001258502.1
NM_003847.2 488 Missense Mutation TCT,TGT S151C NP_003838.1
Gene
PLIN1
Gene Name
perilipin 1
There are no transcripts associated with this gene.

Gene
WDR93
Gene Name
WD repeat domain 93
There are no transcripts associated with this gene.

View Full Product Details