Product Details

SNP ID
rs77487699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11836522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGCCTCAGAATATACTTGACA[A/G]TTGTGGAAACGTTAGCAATCTACTC
Phenotype
MIM: 613746 MIM: 615874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCAR4 PubMed Links

Gene Details

Gene
BCAR4
Gene Name
breast cancer anti-estrogen resistance 4 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RSL1D1
Gene Name
ribosomal L1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015659.2 2811 UTR 3 NP_056474.2

View Full Product Details