Product Details

SNP ID
rs74839932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70662785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATAAAATGACCCCACACCTCC[C/T]ACTGCTCCAGTCACCCCCCGAAAGG
Phenotype
MIM: 612081 MIM: 616951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL34 PubMed Links

Gene Details

Gene
IL34
Gene Name
interleukin 34
There are no transcripts associated with this gene.

Gene
MTSS1L
Gene Name
metastasis suppressor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138383.2 3609 UTR 3 NP_612392.1
XM_005256236.4 3609 UTR 3 XP_005256293.2
XM_011523450.2 3609 UTR 3 XP_011521752.1
XM_011523453.2 3609 UTR 3 XP_011521755.2
XM_011523454.2 3609 UTR 3 XP_011521756.2
XM_011523458.2 3609 UTR 3 XP_011521760.1
XM_017023879.1 3609 UTR 3 XP_016879368.1
XM_017023880.1 3609 UTR 3 XP_016879369.1
XM_017023881.1 3609 UTR 3 XP_016879370.1
XM_017023882.1 3609 Intron XP_016879371.1
XM_017023883.1 3609 Intron XP_016879372.1
XM_017023884.1 3609 Intron XP_016879373.1

View Full Product Details