Product Details
- SNP ID
-
rs78932948
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74953015 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGTCCCTCCTCACCCACTCTGG[C/T]GTTGGGCTCCACTGCCCACTGGCTG
- Phenotype
-
MIM: 605752
MIM: 607828
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HID1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2307010] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HID1
- Gene Name
- HID1 domain containing
- Gene
- OTOP3
- Gene Name
- otopetrin 3
There are no transcripts associated with this gene.
View Full Product Details