Product Details

SNP ID
rs77335949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75635268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGGCGCACAGTAGGGAAGAGT[A/C]AGGCCAGGTCGGGGTCGGGGCACAG
Phenotype
MIM: 603781
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYO15B PubMed Links

Gene Details

Gene
MYO15B
Gene Name
myosin XVB
There are no transcripts associated with this gene.

Gene
RECQL5
Gene Name
RecQ like helicase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003715.3 1246 Intron NP_001003715.1
NM_001003716.3 1246 Intron NP_001003716.1
NM_004259.6 1246 Intron NP_004250.4
XM_005257818.3 1246 Intron XP_005257875.1
XM_005257822.3 1246 Intron XP_005257879.1
XM_005257823.3 1246 Intron XP_005257880.1
XM_006722186.2 1246 Intron XP_006722249.1
XM_011525482.2 1246 Intron XP_011523784.1
XM_011525484.1 1246 Intron XP_011523786.1
XM_011525485.2 1246 Intron XP_011523787.1
XM_011525486.2 1246 Intron XP_011523788.1
XM_017025343.1 1246 Intron XP_016880832.1
XM_017025344.1 1246 Intron XP_016880833.1
Gene
SMIM5
Gene Name
small integral membrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162995.2 1246 Intron NP_001156467.1
XM_011525116.2 1246 UTR 3 XP_011523418.1
XM_011525118.2 1246 Intron XP_011523420.1
XM_011525119.2 1246 Intron XP_011523421.1
XM_017024943.1 1246 Intron XP_016880432.1
XM_017024944.1 1246 Intron XP_016880433.1
XM_017024945.1 1246 Intron XP_016880434.1
XM_017024946.1 1246 Intron XP_016880435.1
XM_017024947.1 1246 Intron XP_016880436.1
XM_017024948.1 1246 Intron XP_016880437.1
Gene
SMIM6
Gene Name
small integral membrane protein 6
There are no transcripts associated with this gene.

View Full Product Details