Product Details

SNP ID
rs74873238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41869532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATCGAGCAGAGGAGTGGCAGGG[A/G]AGTGCTGCCTGACGTAATCTTTGAG
Phenotype
MIM: 108728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACLY PubMed Links

Gene Details

Gene
ACLY
Gene Name
ATP citrate lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001096.2 3163 Missense Mutation TCC,TTC S998F NP_001087.2
NM_001303274.1 3163 Missense Mutation TCC,TTC S1052F NP_001290203.1
NM_001303275.1 3163 Missense Mutation TCC,TTC S1042F NP_001290204.1
NM_198830.1 3163 Missense Mutation TCC,TTC S988F NP_942127.1
XM_005257395.1 3163 Missense Mutation TCC,TTC S998F XP_005257452.1
XM_017024688.1 3163 Missense Mutation TCC,TTC S988F XP_016880177.1
Gene
KLHL11
Gene Name
kelch like family member 11
There are no transcripts associated with this gene.

View Full Product Details