Product Details
- SNP ID
-
rs74509387
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35825395 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCTGTTTTTAAAATCACTTTTAT[G/T]GAGATATCATTTATGTACAATAAAA
- Phenotype
-
MIM: 602716
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NPHS1
PubMed Links
Gene Details
- Gene
- NPHS1
- Gene Name
- NPHS1, nephrin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004646.3 |
5001 |
UTR 3 |
|
|
NP_004637.1 |
View Full Product Details