Product Details

SNP ID
rs79410238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:307349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTTGGGCTGGCGTCTGGCTC[C/T]GGAGCAGTGACAGCTGGCTGGTATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929060 PubMed Links

Gene Details

Gene
LOC101929060
Gene Name
uncharacterized LOC101929060
There are no transcripts associated with this gene.

Gene
MIER2
Gene Name
MIER family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005259582.4 1979 Silent Mutation CCA,CCG P464P XP_005259639.1
XM_006722769.3 1979 Silent Mutation CCA,CCG P457P XP_006722832.1
XM_011528072.2 1979 Silent Mutation CCA,CCG P464P XP_011526374.1
XM_011528073.1 1979 Silent Mutation CCA,CCG P462P XP_011526375.1
XM_011528074.2 1979 Silent Mutation CCA,CCG P457P XP_011526376.1
XM_011528075.2 1979 Silent Mutation CCA,CCG P426P XP_011526377.1
XM_011528077.2 1979 Silent Mutation CCA,CCG P426P XP_011526379.1
XM_011528081.2 1979 Silent Mutation CCA,CCG P386P XP_011526383.1
XM_011528082.2 1979 Silent Mutation CCA,CCG P378P XP_011526384.1

View Full Product Details