Product Details
- SNP ID
-
rs78649129
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11165138 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAAACAGGCCATGAAGGAAACTG[A/T]GAGTGTCTCTGGCAAGCCCCGTTTC
- Phenotype
-
MIM: 614610
MIM: 609394
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KANK2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16423] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KANK2
- Gene Name
- KN motif and ankyrin repeat domains 2
- Gene
- SPC24
- Gene Name
- SPC24, NDC80 kinetochore complex component
There are no transcripts associated with this gene.
View Full Product Details