Product Details

SNP ID
rs78607593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15424398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATTCTAAGGTGGAGAGGGGGAC[C/G]GGAGATGAGTGGGAGGGGTGGATGC
Phenotype
MIM: 609475
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AKAP8L PubMed Links

Gene Details

Gene
AKAP8L
Gene Name
A-kinase anchoring protein 8 like
There are no transcripts associated with this gene.

Gene
WIZ
Gene Name
widely interspaced zinc finger motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021241.2 Intron NP_067064.2
XM_005260004.4 Intron XP_005260061.1
XM_005260005.4 Intron XP_005260062.1
XM_005260006.4 Intron XP_005260063.1
XM_005260007.4 Intron XP_005260064.1
XM_005260008.2 Intron XP_005260065.1
XM_005260009.2 Intron XP_005260066.1
XM_005260010.3 Intron XP_005260067.1
XM_005260011.2 Intron XP_005260068.1
XM_005260012.2 Intron XP_005260069.1
XM_006722828.3 Intron XP_006722891.1
XM_011528163.2 Intron XP_011526465.1

View Full Product Details