Product Details

SNP ID
rs78149184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:666027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCTGGAGTTTGGGATGGACACA[A/G]GGTGAAGTCTGTGTGCCCCTGAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCRT2 PubMed Links

Gene Details

Gene
SCRT2
Gene Name
scratch family transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033129.3 Intron NP_149120.1

View Full Product Details