Product Details

SNP ID
rs75813706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35610652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTTTCCTCTTTGGTAATAAGA[C/T]GTGAATTTGGCATACTAAATTAAAA
Phenotype
MIM: 160000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MB PubMed Links

Gene Details

Gene
MB
Gene Name
myoglobin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005368.2 Intron NP_005359.1
NM_203377.1 Intron NP_976311.1
NM_203378.1 Intron NP_976312.1
XM_005261605.2 Intron XP_005261662.1

View Full Product Details