Product Details

SNP ID
rs75344685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:131105372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGCGAGGAATCGCCGTGGCGT[C/T]TTGGTGTTCTCCACGCTGGTTCGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHB2 PubMed Links

Gene Details

Gene
PLEKHB2
Gene Name
pleckstrin homology domain containing B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100623.1 68 UTR 5 NP_001094093.1
NM_001267062.1 68 UTR 5 NP_001253991.1
NM_001267063.1 68 UTR 5 NP_001253992.1
NM_001267064.1 68 UTR 5 NP_001253993.1
NM_001267065.1 68 UTR 5 NP_001253994.1
NM_001267066.1 68 UTR 5 NP_001253995.1
NM_001267067.1 68 UTR 5 NP_001253996.1
NM_001267068.1 68 UTR 5 NP_001253997.1
NM_001309448.1 68 UTR 5 NP_001296377.1
NM_001309450.1 68 UTR 5 NP_001296379.1
NM_001309451.1 68 UTR 5 NP_001296380.1
NM_001309452.1 68 UTR 5 NP_001296381.1
NM_017958.2 68 UTR 5 NP_060428.2
XM_011511385.2 68 UTR 5 XP_011509687.1
XM_011511386.2 68 UTR 5 XP_011509688.1
XM_017004408.1 68 UTR 5 XP_016859897.1
XM_017004409.1 68 UTR 5 XP_016859898.1
XM_017004410.1 68 UTR 5 XP_016859899.1
XM_017004411.1 68 UTR 5 XP_016859900.1
XM_017004412.1 68 UTR 5 XP_016859901.1
XM_017004413.1 68 UTR 5 XP_016859902.1
Gene
POTEI
Gene Name
POTE ankyrin domain family member I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277406.1 68 Intron NP_001264335.1
XM_011511670.2 68 Intron XP_011509972.1
XM_017004732.1 68 Intron XP_016860221.1
XM_017004733.1 68 Intron XP_016860222.1
XM_017004734.1 68 Intron XP_016860223.1
XM_017004735.1 68 Intron XP_016860224.1

View Full Product Details