Product Details
- SNP ID
-
rs76273111
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:31876107 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATTGTACTTATATAAAGTTCTTA[A/C]CTTCAATTATTACCCTATATTATCA
- Phenotype
-
MIM: 611786
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MEMO1
PubMed Links
Gene Details
- Gene
- MEMO1
- Gene Name
- mediator of cell motility 1
There are no transcripts associated with this gene.
View Full Product Details