Product Details

SNP ID
rs80007105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48375289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGTACAGAAATATTTTAAATT[A/G]TCTTCTTTTGGTTTGGATCATCCCT
Phenotype
MIM: 609075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXW12 PubMed Links

Gene Details

Gene
FBXW12
Gene Name
F-box and WD repeat domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159927.1 Intron NP_001153399.1
NM_001159929.1 Intron NP_001153401.1
NM_207102.2 Intron NP_996985.2
XM_017006224.1 Intron XP_016861713.1

View Full Product Details