Product Details

SNP ID
rs77328224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:168666079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACATCTTCCTCCTTCCTCAGAC[A/G]GGCTATCATTCTTTATTCTTATCCT
Phenotype
MIM: 603745
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLIT3 PubMed Links

Gene Details

Gene
SLIT3
Gene Name
slit guidance ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271946.1 5367 UTR 3 NP_001258875.1
NM_003062.3 5367 UTR 3 NP_003053.1
XM_017009779.1 5367 UTR 3 XP_016865268.1

View Full Product Details