Product Details

SNP ID
rs79449889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160402046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTTCACAGTGTTCTGAGAAGAG[G/T]CTTCAGGATCATGGCAATCTATTAC
Phenotype
MIM: 605974 MIM: 615253
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLU7 PubMed Links

Gene Details

Gene
SLU7
Gene Name
SLU7 homolog, splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006425.4 3502 UTR 3 NP_006416.3
XM_005265793.3 3502 UTR 3 XP_005265850.1
XM_011534418.2 3502 Intron XP_011532720.1
Gene
ZBED8
Gene Name
zinc finger BED-type containing 8
There are no transcripts associated with this gene.

View Full Product Details