Product Details

SNP ID
rs79701050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160403217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAACAAGGATTTTTCTATCTAC[A/G]ATCATCAATAAGAAGCTGAAAAGAA
Phenotype
MIM: 605974 MIM: 615253
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLU7 PubMed Links

Gene Details

Gene
SLU7
Gene Name
SLU7 homolog, splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006425.4 2331 UTR 3 NP_006416.3
XM_005265793.3 2331 UTR 3 XP_005265850.1
XM_011534418.2 2331 UTR 3 XP_011532720.1
Gene
ZBED8
Gene Name
zinc finger BED-type containing 8
There are no transcripts associated with this gene.

View Full Product Details