Product Details

SNP ID
rs75710230
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111302559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGAGGCAGGCACTTAAAAAATA[C/T]CTGCTTTAATCTTTATTCTTTTTAC
Phenotype
MIM: 602776
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
REV3L PubMed Links

Gene Details

Gene
REV3L
Gene Name
REV3 like, DNA directed polymerase zeta catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286431.1 Intron NP_001273360.1
NM_001286432.1 Intron NP_001273361.1
NM_002912.4 Intron NP_002903.3
XM_011536028.1 Intron XP_011534330.1
XM_011536029.2 Intron XP_011534331.1
XM_011536030.2 Intron XP_011534332.1
XM_011536031.2 Intron XP_011534333.1
XM_011536032.2 Intron XP_011534334.1
XM_011536036.2 Intron XP_011534338.1
XM_017011152.1 Intron XP_016866641.1
XM_017011153.1 Intron XP_016866642.1
XM_017011154.1 Intron XP_016866643.1
XM_017011155.1 Intron XP_016866644.1

View Full Product Details