Product Details

SNP ID
rs75653060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137493615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTTGCCAGATGAGAGCGCGCCGC[A/C]CAAGATGGGGTGCACCGGGTGCACG
Phenotype
MIM: 609323
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OLIG3 PubMed Links

Gene Details

Gene
OLIG3
Gene Name
oligodendrocyte transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175747.2 780 Missense Mutation GGC,TGC G186C NP_786923.1

View Full Product Details