Product Details

SNP ID
rs75103878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:153019314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTTCTTCAATTCAGATACATA[C/T]CTTATATTTATTGAAGGATTTATAG
Phenotype
MIM: 607191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS17 PubMed Links

Gene Details

Gene
RGS17
Gene Name
regulator of G-protein signaling 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012419.4 Intron NP_036551.3
XM_017010733.1 Intron XP_016866222.1

View Full Product Details