Product Details

SNP ID
rs77389992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:147587069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCTGTATCTTCTTGCCAATATG[C/T]CTGAAATTAGATCTCAAGATACTAC
Phenotype
MIM: 611197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JAKMIP2 PubMed Links

Gene Details

Gene
JAKMIP2
Gene Name
janus kinase and microtubule interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270934.1 7522 UTR 3 NP_001257863.1
NM_001270941.1 7522 UTR 3 NP_001257870.1
NM_001282282.1 7522 UTR 3 NP_001269211.1
NM_014790.4 7522 UTR 3 NP_055605.2
XM_017010097.1 7522 UTR 3 XP_016865586.1
XM_017010098.1 7522 UTR 3 XP_016865587.1
Gene
JAKMIP2-AS1
Gene Name
JAKMIP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details