Product Details
- SNP ID
-
rs75616585
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:45015534 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTATTTTGCAAACCCTGGTCTA[A/G]GCCCATCTTTACAGAGAAGAGATCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DBX2
PubMed Links
Gene Details
- Gene
- DBX2
- Gene Name
- developing brain homeobox 2
View Full Product Details