Product Details

SNP ID
rs79935822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197005870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCATCCTTAAGAGAGGCCAGA[A/G]GAGAGGACTTGTTAACTCCAGAGCA
Phenotype
MIM: 155750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MELTF PubMed Links

Gene Details

Gene
MELTF
Gene Name
melanotransferrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005929.5 Intron NP_005920.2
NM_033316.3 Intron NP_201573.1
XM_006713643.3 Intron XP_006713706.2
XM_011512850.2 Intron XP_011511152.1
XM_011512851.2 Intron XP_011511153.1
Gene
MFI2-AS1
Gene Name
MFI2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details