Product Details
- SNP ID
-
rs78024984
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:80082155 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTTTGGTTTCAGGTATTGGCTT[G/T]CCTGGCTGATTCATGTGGGAGAGTC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM254
PubMed Links
Gene Details
- Gene
- TMEM254
- Gene Name
- transmembrane protein 254
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001270367.1 |
351 |
Missense Mutation |
GCC,TCC |
A92S |
NP_001257296.1 |
NM_001270368.1 |
351 |
Missense Mutation |
GCC,TCC |
A92S |
NP_001257297.1 |
NM_001270369.1 |
351 |
Missense Mutation |
GCC,TCC |
A92S |
NP_001257298.1 |
NM_001270370.1 |
351 |
Missense Mutation |
GCC,TCC |
A68S |
NP_001257299.1 |
NM_001270371.1 |
351 |
Missense Mutation |
TTG,TTT |
L99F |
NP_001257300.1 |
NM_001270372.1 |
351 |
Missense Mutation |
GCC,TCC |
A68S |
NP_001257301.1 |
NM_001270373.1 |
351 |
Missense Mutation |
GCC,TCC |
A68S |
NP_001257302.1 |
NM_001270374.1 |
351 |
Missense Mutation |
TTG,TTT |
L74F |
NP_001257303.1 |
NM_025125.3 |
351 |
Missense Mutation |
GCC,TCC |
A68S |
NP_079401.2 |
XM_006717978.2 |
351 |
Missense Mutation |
GCC,TCC |
A77S |
XP_006718041.1 |
XM_011540192.2 |
351 |
Missense Mutation |
GCC,TCC |
A92S |
XP_011538494.1 |
XM_011540193.2 |
351 |
Missense Mutation |
TTG,TTT |
L74F |
XP_011538495.1 |
XM_017016679.1 |
351 |
Missense Mutation |
GCC,TCC |
A92S |
XP_016872168.1 |
XM_017016680.1 |
351 |
Missense Mutation |
GCC,TCC |
A77S |
XP_016872169.1 |
- Gene
- TMEM254-AS1
- Gene Name
- TMEM254 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details