Product Details

SNP ID
rs76719640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84195872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGCTGTGTGACCTTGAACAAAT[A/G]TATTTACCTCCCTGAGCCCCAGGTC
Phenotype
MIM: 609502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDHR1 PubMed Links

Gene Details

Gene
CDHR1
Gene Name
cadherin related family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171971.2 Intron NP_001165442.1
NM_033100.3 Intron NP_149091.1
XM_011540337.1 Intron XP_011538639.1
XM_011540338.1 Intron XP_011538640.1
XM_011540339.1 Intron XP_011538641.1
XM_011540340.2 Intron XP_011538642.1

View Full Product Details