Product Details

SNP ID
rs74616092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119499952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTTTATGTTAGCTTAGCCATC[A/T]TGCAAAATTTACTGGTGAAGCAGTT
Phenotype
MIM: 602856
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RGS10 PubMed Links

Gene Details

Gene
RGS10
Gene Name
regulator of G-protein signaling 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005339.1 779 UTR 3 NP_001005339.1
NM_002925.3 779 UTR 3 NP_002916.1
XM_017016492.1 779 UTR 3 XP_016871981.1
XM_017016493.1 779 UTR 3 XP_016871982.1

View Full Product Details