Product Details

SNP ID
rs76367688
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:85265856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGTGTTAAAACTTATCTATAT[A/G]TAGGTGTTATTGTATATCTAAATTA
Phenotype
MIM: 603517
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL10 PubMed Links

Gene Details

Gene
BCL10
Gene Name
B-cell CLL/lymphoma 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320715.1 3025 UTR 3 NP_001307644.1
NM_003921.4 3025 UTR 3 NP_003912.1
XM_011542397.2 3025 Intron XP_011540699.1
XM_011542398.2 3025 Intron XP_011540700.1
XM_011542399.2 3025 Intron XP_011540701.1
Gene
C1orf52
Gene Name
chromosome 1 open reading frame 52
There are no transcripts associated with this gene.

View Full Product Details