Product Details

SNP ID
rs79613472
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:109476793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCAGGTTGCACCGGATCCAAT[A/T]TGAGATGCCCCTCTGCGATGAAGAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATXN7L2 PubMed Links

Gene Details

Gene
ATXN7L2
Gene Name
ataxin 7 like 2
There are no transcripts associated with this gene.

Gene
SYPL2
Gene Name
synaptophysin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040709.1 638 Missense Mutation TAT,TTT Y91F NP_001035799.1
XM_011541283.2 638 Missense Mutation TAT,TTT Y91F XP_011539585.1
XM_011541284.2 638 Missense Mutation TAT,TTT Y91F XP_011539586.1
XM_011541285.1 638 Missense Mutation TAT,TTT Y91F XP_011539587.1

View Full Product Details