Product Details

SNP ID
rs80338137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113798637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACTGAGTATGAGGTTTTCTGG[A/T]TTAAAGATAGAAAAGAAAGGAGAGA
Phenotype
MIM: 610748
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC390251 PubMed Links

Gene Details

Gene
LOC390251
Gene Name
SH3 domain containing GRB2 like 1 pseudogene
There are no transcripts associated with this gene.

Gene
USP28
Gene Name
ubiquitin specific peptidase 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301029.1 4091 UTR 3 NP_001287958.1
NM_020886.3 4091 UTR 3 NP_065937.1
XM_005271630.2 4091 UTR 3 XP_005271687.1
XM_005271631.2 4091 UTR 3 XP_005271688.1
XM_005271632.2 4091 UTR 3 XP_005271689.1
XM_005271633.4 4091 UTR 3 XP_005271690.1
XM_005271636.2 4091 UTR 3 XP_005271693.1
XM_005271637.2 4091 UTR 3 XP_005271694.1
XM_005271638.2 4091 UTR 3 XP_005271695.1
XM_005271639.2 4091 UTR 3 XP_005271696.1
XM_011542936.2 4091 UTR 3 XP_011541238.1
XM_011542938.1 4091 UTR 3 XP_011541240.1
XM_011542941.1 4091 UTR 3 XP_011541243.1
XM_011542942.1 4091 UTR 3 XP_011541244.1
XM_017018056.1 4091 UTR 3 XP_016873545.1
XM_017018057.1 4091 UTR 3 XP_016873546.1
XM_017018058.1 4091 UTR 3 XP_016873547.1
XM_017018059.1 4091 UTR 3 XP_016873548.1
XM_017018060.1 4091 UTR 3 XP_016873549.1
XM_017018061.1 4091 UTR 3 XP_016873550.1
XM_017018062.1 4091 UTR 3 XP_016873551.1
XM_017018063.1 4091 UTR 3 XP_016873552.1
XM_017018064.1 4091 UTR 3 XP_016873553.1
XM_017018065.1 4091 UTR 3 XP_016873554.1
XM_017018066.1 4091 UTR 3 XP_016873555.1
XM_017018067.1 4091 UTR 3 XP_016873556.1
XM_017018068.1 4091 UTR 3 XP_016873557.1
XM_017018069.1 4091 Intron XP_016873558.1

View Full Product Details