Product Details

SNP ID
rs76795438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123976705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTGATAGCACAGACTATGGGGG[G/T]GGGCTGCCTGCTGTAGACTCTCGGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10S1 PubMed Links

Gene Details

Gene
OR10S1
Gene Name
olfactory receptor family 10 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004474.1 987 Silent Mutation CCA,CCC P329P NP_001004474.1

View Full Product Details