Product Details

SNP ID
rs78568067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124424353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAACGTCACACAGATAATGGTC[A/C]ATGACGTTGGAATCACAGAAGGTCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR8B4 PubMed Links

Gene Details

Gene
OR8B4
Gene Name
olfactory receptor family 8 subfamily B member 4 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005196.1 519 Missense Mutation ATG,ATT M173I NP_001005196.1

View Full Product Details