Product Details

SNP ID
rs75988347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124920800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGAGCACAGGATAGTCACGGG[G/T]GTTAGGTTACTGATGTTAGTTTCCA
Phenotype
MIM: 611642 MIM: 611641
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HEPACAM PubMed Links

Gene Details

Gene
HEPACAM
Gene Name
hepatic and glial cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152722.4 1992 UTR 3 NP_689935.2
XM_005271449.2 1992 UTR 3 XP_005271506.1
XM_017017361.1 1992 UTR 3 XP_016872850.1
Gene
HEPN1
Gene Name
hepatocellular carcinoma, down-regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037558.2 1992 Intron NP_001032647.2

View Full Product Details