Product Details

SNP ID
rs76744140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125675828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACAGCTAGTCTCTCTGCCTTC[C/T]CTTAGGCCTAATGGGAAGGGCATCT
Phenotype
MIM: 102525 MIM: 603078
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACRV1 PubMed Links

Gene Details

Gene
ACRV1
Gene Name
acrosomal vesicle protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001612.5 Intron NP_001603.1
NM_020069.4 Intron NP_064454.1
NM_020107.4 Intron NP_064492.1
NM_020108.4 Intron NP_064493.1
Gene
CHEK1
Gene Name
checkpoint kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114121.2 Intron NP_001107593.1
NM_001114122.2 Intron NP_001107594.1
NM_001244846.1 Intron NP_001231775.1
NM_001274.5 Intron NP_001265.2
XM_011542560.2 Intron XP_011540862.1
XM_011542561.2 Intron XP_011540863.1
XM_011542562.2 Intron XP_011540864.1
XM_011542563.2 Intron XP_011540865.1
XM_017017146.1 Intron XP_016872635.1

View Full Product Details