Product Details

SNP ID
rs79613010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:610079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGGATGAAGGTCTCATTGCAAG[C/T]GCTGTGCATCAATGTCATCCGTGCA
Phenotype
MIM: 180072
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE6B PubMed Links

Gene Details

Gene
PDE6B
Gene Name
phosphodiesterase 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000283.3 Intron NP_000274.2
NM_001145291.1 Intron NP_001138763.1
NM_001145292.1 Intron NP_001138764.1
XM_011513473.2 Intron XP_011511775.1
XM_011513474.2 Intron XP_011511776.1
XM_011513475.2 Intron XP_011511777.1
XM_011513476.2 Intron XP_011511778.1
XM_011513477.2 Intron XP_011511779.1
XM_011513478.2 Intron XP_011511780.1
XM_017008283.1 Intron XP_016863772.1
XM_017008284.1 Intron XP_016863773.1
XM_017008285.1 Intron XP_016863774.1
XM_017008286.1 Intron XP_016863775.1
XM_017008287.1 Intron XP_016863776.1
XM_017008288.1 Intron XP_016863777.1
XM_017008289.1 Intron XP_016863778.1

View Full Product Details