Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_030792.6 | Intron | NP_110419.5 | |||
XM_006718697.3 | Intron | XP_006718760.1 | |||
XM_011545276.2 | Intron | XP_011543578.1 | |||
XM_011545277.2 | Intron | XP_011543579.1 | |||
XM_011545278.2 | Intron | XP_011543580.1 | |||
XM_011545279.2 | Intron | XP_011543581.1 | |||
XM_011545280.1 | Intron | XP_011543582.1 | |||
XM_011545281.2 | Intron | XP_011543583.1 | |||
XM_011545285.2 | Intron | XP_011543587.1 | |||
XM_011545286.1 | Intron | XP_011543588.1 | |||
XM_011545287.1 | Intron | XP_011543589.1 |