Product Details

SNP ID
rs117330960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227330927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAACTTTTGAGTTTTTCTAAA[A/T]GTTATTTAGAAAAGGAGTTGGAAAA
Phenotype
MIM: 614785
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC654841 PubMed Links

Gene Details

Gene
LOC654841
Gene Name
uncharacterized LOC654841
There are no transcripts associated with this gene.

Gene
MFF
Gene Name
mitochondrial fission factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277061.1 Intron NP_001263990.1
NM_001277062.1 Intron NP_001263991.1
NM_001277063.1 Intron NP_001263992.1
NM_001277064.1 Intron NP_001263993.1
NM_001277065.1 Intron NP_001263994.1
NM_001277066.1 Intron NP_001263995.1
NM_001277067.1 Intron NP_001263996.1
NM_001277068.1 Intron NP_001263997.1
NM_020194.5 Intron NP_064579.3
XM_005246681.3 Intron XP_005246738.1
XM_005246682.1 Intron XP_005246739.1
XM_005246683.1 Intron XP_005246740.1
XM_005246684.1 Intron XP_005246741.1
XM_005246685.3 Intron XP_005246742.1
XM_005246686.3 Intron XP_005246743.1
XM_005246687.3 Intron XP_005246744.1
XM_005246688.1 Intron XP_005246745.1
XM_006712637.1 Intron XP_006712700.1
XM_006712638.1 Intron XP_006712701.1
XM_006712639.2 Intron XP_006712702.1
XM_011511500.1 Intron XP_011509802.1
XM_011511501.1 Intron XP_011509803.1
XM_017004512.1 Intron XP_016860001.1
XM_017004513.1 Intron XP_016860002.1
XM_017004514.1 Intron XP_016860003.1
XM_017004515.1 Intron XP_016860004.1

View Full Product Details