Product Details
- SNP ID
-
rs115376825
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:231976183 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGGCTTTTAGTGTACCCATCACC[C/T]CTATAGTGTACATTACAACTCTTTT
- Phenotype
-
MIM: 614184
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DIS3L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34169389] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DIS3L2
- Gene Name
- DIS3 like 3'-5' exoribonuclease 2
View Full Product Details