Product Details

SNP ID
rs115073548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129315687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAACCACGGAACCTTCTTGGCCTC[A/G]GTGTAGATCTTGGCCAGCGACGAGC
Phenotype
MIM: 602785
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
H1FX PubMed Links

Gene Details

Gene
H1FX
Gene Name
H1 histone family member X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006026.3 591 Silent Mutation ACC,ACT T72T NP_006017.1
Gene
H1FX-AS1
Gene Name
H1FX antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details