Product Details

SNP ID
rs116187706
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1716389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCCCCTTCTGAGCCTGGGAGA[A/C]CCTAGGGGAGTGGGTAGCCTGGGCT
Phenotype
MIM: 602422 MIM: 605303 MIM: 615975
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
SLBP PubMed Links

Gene Details

Gene
SLBP
Gene Name
stem-loop binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306074.1 2400 Intron NP_001293003.1
NM_001306075.1 2400 Intron NP_001293004.1
NM_006527.3 2400 Intron NP_006518.1
Gene
TACC3
Gene Name
transforming acidic coiled-coil containing protein 3
There are no transcripts associated with this gene.

Gene
TMEM129
Gene Name
transmembrane protein 129
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127266.1 2400 UTR 3 NP_001120738.1
NM_138385.3 2400 UTR 3 NP_612394.1
XM_005248038.4 2400 UTR 3 XP_005248095.1
XM_005248039.4 2400 UTR 3 XP_005248096.1
XM_011513594.2 2400 UTR 3 XP_011511896.1
XM_017008834.1 2400 UTR 3 XP_016864323.1

View Full Product Details