Product Details

SNP ID
rs111252689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17843353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAATGAAGATGAGACTACCAAG[A/G]CGAGCCAAAACCGCAGCACTAGAAA
Phenotype
MIM: 611799 MIM: 606280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCORL PubMed Links

Gene Details

Gene
LCORL
Gene Name
ligand dependent nuclear receptor corepressor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166139.2 8180 Intron NP_001159611.1
NM_153686.8 8180 UTR 3 NP_710153.2
XM_011513821.2 8180 UTR 3 XP_011512123.1
XM_011513822.2 8180 UTR 3 XP_011512124.1
XM_011513823.2 8180 UTR 3 XP_011512125.1
XM_011513825.2 8180 Intron XP_011512127.1
XM_011513826.2 8180 UTR 3 XP_011512128.1
XM_017007961.1 8180 UTR 3 XP_016863450.1
XM_017007962.1 8180 UTR 3 XP_016863451.1
XM_017007963.1 8180 UTR 3 XP_016863452.1
XM_017007964.1 8180 Intron XP_016863453.1
XM_017007965.1 8180 Intron XP_016863454.1
Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022346.4 8180 Silent Mutation AGA,AGG R992R NP_071741.2
XM_011513876.2 8180 Silent Mutation AGA,AGG R763R XP_011512178.1
XM_017008543.1 8180 Silent Mutation AGA,AGG R994R XP_016864032.1
XM_017008544.1 8180 Silent Mutation AGA,AGG R765R XP_016864033.1

View Full Product Details