Product Details

SNP ID
rs112435588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:31194670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTCCACGGCTGGCCAGGCTCA[A/T]ATCTGGGGGATAAAGCCTTCCCCGG
Phenotype
MIM: 603007
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDH6 PubMed Links

Gene Details

Gene
CDH6
Gene Name
cadherin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004932.3 Intron NP_004923.1
XM_011513921.2 Intron XP_011512223.1
XM_017008910.1 Intron XP_016864399.1
XM_017008911.1 Intron XP_016864400.1

View Full Product Details