Product Details

SNP ID
rs113636604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55231547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACGGGCCGCGAGACCCGCAGC[A/G]TGCGGTCCGCCAGCGCCAGGCAGCA
Phenotype
MIM: 607752 MIM: 614086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNO PubMed Links

Gene Details

Gene
CCNO
Gene Name
cyclin O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021147.4 1075 Missense Mutation ACG,ATG T294M NP_066970.3
Gene
MCIDAS
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
There are no transcripts associated with this gene.

View Full Product Details