Product Details

SNP ID
rs117431205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92123079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAAAATTAGCCACAGATCCTC[A/C]AAGTCATAAGGCAGCAATGAAACTG
Phenotype
MIM: 601637
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYP51A1 PubMed Links

Gene Details

Gene
CYP51A1
Gene Name
cytochrome P450 family 51 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000786.3 Intron NP_000777.1
NM_001146152.1 Intron NP_001139624.1

View Full Product Details